Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrazol-3-ol 97%

Supplier: AMBEED, INC
Warning

A125115-100G A125115-10G A125115-1G A125115-250MG A125115-25G A125115-5G
76810-796EA 295 USD InStock
76810-796 76810-798 76810-800 76810-802 76810-804 76810-806
1H-Pyrazol-3-ol 97%
1H-Pyrazol-3-ol
Precaución: for Research Use Only


Formula: C₃H₄N₂O
MDL Number: MFCD20527386
CAS Number: 60456-92-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty