Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-5-chloropyridin-2-amine 97%

Supplier: AMBEED, INC
Warning

A125200-100MG A125200-10G A125200-1G A125200-250MG A125200-25G A125200-5G
76810-822EA 5 USD InStock
76810-822 76810-824 76810-826 76810-828 76810-830 76810-832
6-Bromo-5-chloropyridin-2-amine 97%
6-Bromo-5-chloropyridin-2-amine
Precaución: for Research Use Only


Formula: C₅H₄BrClN₂
MDL Number: MFCD16556292
CAS Number: 1004294-58-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty