Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium[[(tert-butoxycarbonyl)amino]methyl]trifluoroborate 95%

Supplier: AMBEED, INC
Warning

A139380-100MG A139380-1G A139380-250MG A139380-5G
76811-700EA 5 USD InStock
76811-700 76811-702 76811-704 77549-692
Potassium[[(tert-butoxycarbonyl)amino]methyl]trifluoroborate 95%
Potassium[[(tert-butoxycarbonyl)amino]methyl]trifluoroborate
Precaución: for Research Use Only


Formula: C₆H₁₂BF₃KNO₂
MDL Number: MFCD19686142
CAS Number: 1314538-55-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty