Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-6-methylpyrimidin-4-ol 97%

Supplier: AMBEED, INC
Warning

A141074-100MG A141074-1G A141074-250MG A141074-5G
76811-776EA 8 USD InStock
76811-776 76811-778 76811-780 77549-716
5-Bromo-6-methylpyrimidin-4-ol 97%
5-Bromo-6-methylpyrimidin-4-ol
Precaución: for Research Use Only


Formula: C₅H₅BrN₂O
MDL Number: MFCD01646062
CAS Number: 3438-52-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty