Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (2,6-dichloropyridin-4-yl)carbamate 95%

Supplier: AMBEED, INC
Warning

A154234-100MG A154234-1G A154234-25G A154234-10G
76812-384EA 8 USD InStock
76812-384 76812-386 76812-388 77671-172
tert-Butyl (2,6-dichloropyridin-4-yl)carbamate 95%
tert-Butyl (2,6-dichloropyridin-4-yl)carbamate
Precaución: for Research Use Only


Formula: C₁₀H₁₂Cl₂N₂O₂
MDL Number: MFCD03070211
CAS Number: 501907-61-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty