Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Methoxy-2-(trifluoromethyl)-4H-chromen-4-one 97%

Supplier: AMBEED, INC
Warning

A203702-1G A203702-250MG
76814-374EA 29 USD InStock
76814-374 76814-376
7-Methoxy-2-(trifluoromethyl)-4H-chromen-4-one 97%
7-Methoxy-2-(trifluoromethyl)-4H-chromen-4-one
Precaución: for Research Use Only


Formula: C₁₁H₇F₃O₃
MDL Number: MFCD00760804
CAS Number: 578-84-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty