Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3-Oxocyclopentanecarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A212079-1G A212079-250MG A212079-5G A212079-100MG
76814-576EA 45 USD InStock
76814-576 76814-578 76814-580 77671-890
(R)-3-Oxocyclopentanecarboxylic acid 97%
(R)-3-Oxocyclopentanecarboxylic acid
Precaución: for Research Use Only


Formula: C₆H₈O₃
MDL Number: MFCD19228224
CAS Number: 13012-38-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty