Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-oxotetrahydro-2H-pyran-3-carboxylate 93% (total of isomer)

Supplier: AMBEED, INC
Warning

A227280-1G A227280-250MG
76815-072EA 161 USD InStock
76815-072 76815-074
Ethyl-4-oxotetrahydro-2H-pyran-3-carboxylate 93% (total of isomer)
Ethyl-4-oxotetrahydro-2H-pyran-3-carboxylate
Precaución: for Research Use Only


Formula: C₈H₁₂O₄
MDL Number: MFCD13190006
CAS Number: 141419-94-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty