Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Oxetane-3-carbaldehyde 98%

Supplier: AMBEED, INC
Warning

A332720-100MG A332720-250MG
76817-186EA 31 USD InStock
76817-186 76817-188
Oxetane-3-carbaldehyde 98%
Oxetane-3-carbaldehyde
Precaución: for Research Use Only


Formula: C₄H₆O₂
MDL Number: MFCD21604173
CAS Number: 1305207-52-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty