Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,6,7,8-Tetrahydro-1,6-naphthyridine dihydrochloride 97%

Supplier: AMBEED, INC
Warning

A357704-100MG A357704-1G A357704-250MG
76817-382EA 28 USD InStock
76817-382 76817-384 76817-386
5,6,7,8-Tetrahydro-1,6-naphthyridine dihydrochloride 97%
5,6,7,8-Tetrahydro-1,6-naphthyridine dihydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₂Cl₂N₂
MDL Number: MFCD09997813
CAS Number: 348623-30-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty