Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Borono-2,6-difluorobenzoic acid 96%

Supplier: AMBEED, INC
Warning

A367081-1G A367081-250MG A367081-5G
76817-464EA 61 USD InStock
76817-464 76817-466 76817-468
4-Borono-2,6-difluorobenzoic acid 96%
4-Borono-2,6-difluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₅BF₂O₄
MDL Number: MFCD13181636
CAS Number: 1029716-94-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty