Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3S,4R)-1-tert-Butyl 3-ethyl 4-aminopyrrolidine-1,3-dicarboxylate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A485932-100MG A485932-250MG
76818-644EA 152 USD InStock
76818-644 76818-646
(3S,4R)-1-tert-Butyl 3-ethyl 4-aminopyrrolidine-1,3-dicarboxylate hydrochloride 95%
(3S,4R)-1-tert-Butyl 3-ethyl 4-aminopyrrolidine-1,3-dicarboxylate hydrochloride
Precaución: For research use only.


Formula: C₁₂H₂₃ClN₂O₄
MDL Number: MFCD21608674
CAS Number: 955138-54-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty