Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-2-(2,3-dihydro-1H-inden-1-yl)acetic acid 95%

Supplier: AMBEED, INC
Warning

A694074-100MG A694074-250MG
76820-002EA 74 USD InStock
76820-002 76820-004
2-Amino-2-(2,3-dihydro-1H-inden-1-yl)acetic acid 95%
2-Amino-2-(2,3-dihydro-1H-inden-1-yl)acetic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₃NO₂
MDL Number: MFCD20658678
CAS Number: 179185-78-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty