Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Bis((S)-((1S,2R,4S,5R)-5-ethylquinuclidin-2-yl)(6-methoxyquinolin-4-yl)methoxy)phthalazine 99+%

Supplier: AMBEED, INC
Warning

A785708-100G A785708-10G A785708-1G A785708-250MG A785708-25G A785708-5G
76820-474EA 1035 USD InStock
76820-474 76820-476 76820-478 76820-480 76820-482 76820-484
1,4-Bis((S)-((1S,2R,4S,5R)-5-ethylquinuclidin-2-yl)(6-methoxyquinolin-4-yl)methoxy)phthalazine 99+%
1,4-Bis((S)-((1S,2R,4S,5R)-5-ethylquinuclidin-2-yl)(6-methoxyquinolin-4-yl)methoxy)phthalazine
Precaución: for Research Use Only


Formula: C₄₈H₅₄N₆O₄
MDL Number: MFCD00198107
CAS Number: 140853-10-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty