Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,6-Dichloropyrazine-2-carbonitrile 98%

Supplier: AMBEED, INC
Warning

A119081-100G A119081-100MG A119081-10G A119081-1G A119081-250MG A119081-25G A119081-5G
76822-298EA 264 USD InStock
76822-298 76822-300 76822-302 76822-304 76822-306 76822-308 76822-310
3,6-Dichloropyrazine-2-carbonitrile 98%
3,6-Dichloropyrazine-2-carbonitrile
Precaución: for Research Use Only


Formula: C₅HCl₂N₃
MDL Number: MFCD22199269
CAS Number: 356783-16-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty