Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-6-(1-methyl-1H-pyrazol-4-yl)pyridazine 95%

Supplier: AMBEED, INC
Warning

A130384-1G A130384-250MG A130384-5G
76822-996EA 57 USD InStock
76822-996 76822-998 76823-000
3-Chloro-6-(1-methyl-1H-pyrazol-4-yl)pyridazine 95%
3-Chloro-6-(1-methyl-1H-pyrazol-4-yl)pyridazine
Precaución: for Research Use Only


Formula: C₈H₇ClN₄
MDL Number: MFCD16109166
CAS Number: 943541-20-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty