Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(tert-Butyl)-2-iodophenol 97%

Supplier: AMBEED, INC
Warning

A139872-1G A139872-250MG A139872-5G
76823-380EA 30 USD InStock
76823-380 76823-382 76823-384
4-(tert-Butyl)-2-iodophenol 97%
4-(tert-Butyl)-2-iodophenol
Precaución: for Research Use Only


Formula: C₁₀H₁₃IO
MDL Number: MFCD11035758
CAS Number: 38941-98-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty