Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-1H-pyrazole 97%

Supplier: AMBEED, INC
Warning

A140011-100MG A140011-1G A140011-250MG A140011-5G
76823-390EA 23 USD InStock
76823-390 76823-392 76823-394 76823-396
5-Chloro-1H-pyrazole 97%
5-Chloro-1H-pyrazole
Precaución: for Research Use Only


Formula: C₃H₃ClN₂
MDL Number: MFCD25980930
CAS Number: 934758-92-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty