Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-chloro-3-methylpyrazine 98%

Supplier: AMBEED, INC
Danger

A183807-100MG A183807-1G A183807-250MG
76825-020EA 39 USD InStock
76825-020 76825-022 76825-024
2-Bromo-5-chloro-3-methylpyrazine 98%
2-Bromo-5-chloro-3-methylpyrazine
Precaución: for Research Use Only


Formula: C₅H₄BrClN₂
MDL Number: MFCD18250174
CAS Number: 1260664-82-1
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty