Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6,8-Dibromo-[1,2,4]triazolo[1,5-a]pyridine 97%

Supplier: AMBEED, INC
Warning

A241045-1G A241045-250MG
76826-718EA 43 USD InStock
76826-718 76826-720
6,8-Dibromo-[1,2,4]triazolo[1,5-a]pyridine 97%
6,8-Dibromo-[1,2,4]triazolo[1,5-a]pyridine
Precaución: for Research Use Only


Formula: C₆H₃Br₂N₃
MDL Number: MFCD22124121
CAS Number: 1310680-10-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty