Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bicyclo[2.2.2]octane-1,4-diyldimethanol 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A357021-1G A357021-250MG A357021-5G A357021-25G
76828-690EA 31 USD InStock
76828-690 76828-692 76828-694 77684-050
Bicyclo[2.2.2]octane-1,4-diyldimethanol 98%, Ambeed.Inc
Bicyclo[2.2.2]octane-1,4-diyldimethanol
Precaución: For research use only.


Formula: C₁₀H₁₈O₂
MDL Number: MFCD23135544
CAS Number: 826-45-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty