Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R)-2-(Propan-2-yl)morpholine 98%

Supplier: AMBEED, INC
Danger

A632601-100MG A632601-1G A632601-250MG
76831-180EA 12 USD InStock
76831-180 76831-182 76831-184
(2R)-2-(Propan-2-yl)morpholine 98%
(2R)-2-(Propan-2-yl)morpholine
Precaución: for Research Use Only


Formula: C₇H₁₅NO
MDL Number: MFCD14582703
CAS Number: 792886-64-7
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty