Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Pyrrolidin-1-yl)pyridine-2-carbonitrile 98%

Supplier: AMBEED, INC
Warning

A708711-10G A708711-1G A708711-250MG A708711-5G
76831-688EA 471 USD InStock
76831-688 76831-690 76831-692 76831-694
4-(Pyrrolidin-1-yl)pyridine-2-carbonitrile 98%
4-(Pyrrolidin-1-yl)pyridine-2-carbonitrile
Precaución: for Research Use Only


Formula: C₁₀H₁₁N₃
MDL Number: MFCD08062860
CAS Number: 127680-86-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty