Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo[b]thiophene-3-boronic acid pinacol ester ≥98%

Supplier: AMBEED, INC
Warning

A776121-1G A776121-5G
76832-106EA 11 USD InStock
76832-106 76832-108
Benzo[b]thiophene-3-boronic acid pinacol ester ≥98%
Benzo[b]thiophene-3-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₁₇BO₂S
MDL Number: MFCD11110552
CAS Number: 171364-86-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty