Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Bromoethyl)diphenylsulfonium trifluoromethanesulfonate 95%

Supplier: AMBEED, INC
Warning

A997162-1G A997162-250MG A997162-5G
76833-160EA 25 USD InStock
76833-160 76833-162 76833-164
(2-Bromoethyl)diphenylsulfonium trifluoromethanesulfonate 95%
(2-Bromoethyl)diphenylsulfonium trifluoromethanesulfonate
Precaución: for Research Use Only


Formula: C₁₅H₁₄BrF₃O₃S₂
MDL Number: MFCD12546049
CAS Number: 247129-85-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty