Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-(5-bromopyrimidin-4-yl)acetate 97%

Supplier: AMBEED, INC
Warning

A760749-1G A760749-100MG A760749-250MG A760749-5G
76845-114EA 363 USD InStock
76845-114 76981-416 76981-418 77674-088
Ethyl 2-(5-bromopyrimidin-4-yl)acetate 97%
Ethyl 2-(5-bromopyrimidin-4-yl)acetate
Precaución: For research use only


Formula: C₈H₉BrN₂O₂
MW: 245.07 g/mol
MDL Number: MFCD18642505
CAS Number: 185030-22-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty