Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-8-quinolinol 98%

Supplier: AMBEED, INC
Warning

A106027-1000G A106027-100G A106027-25G A106027-500G
76869-432EA 271 USD InStock
76869-432 76869-434 76869-436 76869-438
2-Methyl-8-quinolinol 98%
2-Methyl-8-quinolinol
Precaución: for Research Use Only


Formula: C₁₀H₉NO
MW: 159.19 g/mol
Boiling Pt: 267
MDL Number: MFCD00006765
CAS Number: 826-81-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty