Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-methoxybenzotrifluoride 97%

Supplier: AMBEED, INC
Warning

A181160-250MG A181160-25G A181160-5G
76872-752EA 14 USD InStock
76872-752 76872-754 76872-756
2-Bromo-4-methoxybenzotrifluoride 97%
2-Bromo-4-methoxybenzotrifluoride
Precaución: for Research Use Only


Formula: C₈H₆BrF₃O
MDL Number: MFCD10696314
CAS Number: 944901-07-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty