Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxy-4-methyl-1H-1,2,4-triazol-5(4h)-one 97%

Supplier: AMBEED, INC
Warning

A191085-10G A191085-1G A191085-250MG A191085-25G A191085-5G
76873-064EA 69 USD InStock
76873-064 76873-066 76873-068 76873-070 76873-072
3-Methoxy-4-methyl-1H-1,2,4-triazol-5(4h)-one 97%
3-Methoxy-4-methyl-1H-1,2,4-triazol-5(4h)-one
Precaución: for Research Use Only


Formula: C₄H₇N₃O₂
MDL Number: MFCD09744038
CAS Number: 135302-13-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty