Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)propanoic acid 98%

Supplier: AMBEED, INC
Warning

A272700-100MG A272700-1G A272700-250MG
76875-100EA 39 USD InStock
76875-100 76875-102 76875-104
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)propanoic acid 98%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)propanoic acid
Precaución: for Research Use Only


Formula: C₉H₁₇BO₄
MW: 200.04 g/mol
MDL Number: MFCD18914529
CAS Number: 1191063-90-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty