Methyl-4-piperidinecarboxylate hydrochloride 98%
Supplier: AMBEED, INC
|
Warning
|
A280307-10G
A280307-25G
A280307-5G
76875-286EA
7
USD
76875-286
76875-288
76875-290
Methyl-4-piperidinecarboxylate hydrochloride 98%
Methyl 4-piperidinecarboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₄ClNO₂ MW: 179.6446 g/mol |
MDL Number:
MFCD02667641 CAS Number: 7462-86-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...