Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A394512-100MG A394512-1G A394512-250MG A394512-5G
76876-826EA 36 USD InStock
76876-826 76876-828 76876-830 76876-832
2-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid 98%
2-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₅BFNO₃
MDL Number: MFCD08436068
CAS Number: 874289-43-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty