Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Carbamoyl-5-fluorobenzeneboronic acid 98%

Supplier: AMBEED, INC
Warning

A441434-100MG A441434-1G A441434-250MG A441434-5G
76877-378EA 37 USD InStock
76877-378 76877-380 76877-382 76877-384
3-Carbamoyl-5-fluorobenzeneboronic acid 98%
3-Carbamoyl-5-fluorobenzeneboronic acid
Precaución: for Research Use Only


Formula: C₇H₇BFNO₃
MDL Number: MFCD08056357
CAS Number: 871332-66-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty