Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-(methylsulfonyl)benzoic acid 98%

Supplier: AMBEED, INC
Warning

A461628-100MG A461628-1G A461628-250MG
76877-610EA 16 USD InStock
76877-610 76877-612 76877-614
2-Fluoro-5-(methylsulfonyl)benzoic acid 98%
2-Fluoro-5-(methylsulfonyl)benzoic acid
Precaución: for Research Use Only


Formula: C₈H₇FO₄S
MDL Number: MFCD11179900
CAS Number: 247569-56-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty