Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Fluoro-2-methylquinoline-8-boronic acid 96%

Supplier: AMBEED, INC
Danger

A604075-100MG A604075-1G A604075-250MG
76878-954EA 39 USD InStock
76878-954 76878-956 76878-958
7-Fluoro-2-methylquinoline-8-boronic acid 96%
7-Fluoro-2-methylquinoline-8-boronic acid
Precaución: for Research Use Only


Formula: C₁₀H₉BFNO₂
MDL Number: MFCD11504838
CAS Number: 1072945-61-9
UN: 3259
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty