Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-naphthoic acid 97%

Supplier: AMBEED, INC
Warning

A722167-100MG A722167-1G A722167-250MG A722167-5G
76879-816EA 44 USD InStock
76879-816 76879-818 76879-820 76879-822
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-naphthoic acid 97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-naphthoic acid
Precaución: for Research Use Only


Formula: C₁₇H₁₉BO₄
MDL Number: MFCD09972180
CAS Number: 1073353-77-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty