Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Bromo-2-methoxyphenyl)-1,3-dioxolane 95%

Supplier: AOB CHEM USA
Warning

16447-1G 16447-25G 16447-5G 16447-500MG
76883-382EA 108.22 USD InStock
76883-382 76883-384 76883-386 77644-748
2-(3-Bromo-2-methoxyphenyl)-1,3-dioxolane 95%
2-(3-Bromo-2-methoxyphenyl)-1,3-dioxolane


Formula: C₁₀H₁₁BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD31805739
CAS Number: 2271442-83-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty