Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,4-trichlorobenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

34068-1G 34068-25G 34068-500MG 34068-5G
76887-734EA 265.23 USD InStock
76887-734 76887-736 76887-738 76887-740
2,3,4-trichlorobenzoic acid ≥97%
2,3,4-trichlorobenzoic acid


Formula: C₇H₃Cl₃O₂
Storage Temperature: Refrigerator
CAS Number: 50-75-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty