Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-methyl 3-(3-chloro-4-methoxyphenyl)acrylate ≥97%

Supplier: AOB CHEM USA
Warning

38114-1G 38114-25G 38114-500MG 38114-5G
76889-338EA 361.77 USD InStock
76889-338 76889-340 76889-342 76889-344
(E)-methyl 3-(3-chloro-4-methoxyphenyl)acrylate ≥97%
(E)-methyl 3-(3-chloro-4-methoxyphenyl)acrylate


Formula: C₁₁H₁₁ClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD20440906
CAS Number: 1187248-43-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty