Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-(benzyloxy)-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOB CHEM USA
Warning

48378-1G 48378-25G 48378-500MG 48378-5G
76890-476EA 129.43 USD InStock
76890-476 76890-478 76890-480 76890-482
2-(2-(benzyloxy)-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2-(benzyloxy)-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane


Formula: C₁₉H₂₂BFO₃
Storage Temperature: Ambient
MDL Number: MFCD23098799
CAS Number: 2246670-77-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty