Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Fluoro-3-iodo-5-methoxybenzene ≥95% research grade

Supplier: AOB CHEM USA
Warning

53109-1G 53109-25G 53109-500MG 53109-5G
76891-362EA 185.66 USD InStock
76891-362 76891-364 76891-366 76891-368
1-Fluoro-3-iodo-5-methoxybenzene ≥95% research grade
1-Fluoro-3-iodo-5-methoxybenzene


Formula: C₇H₆FIO
Storage Temperature: Ambient
MDL Number: MFCD12405142
CAS Number: 1167056-58-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty