Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Isopropylphenyl)methanol ≥97% research grade

Supplier: AOB CHEM USA
Warning

57152-1G 57152-25G 57152-500MG 57152-5G
76891-968EA 203.7 USD InStock
76891-968 76891-970 76891-972 76891-974
(2-Isopropylphenyl)methanol ≥97% research grade
(2-Isopropylphenyl)methanol


Formula: C₁₀H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD9999704
CAS Number: 21190-34-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty