Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-chloro-4-pyridineboronic acid ≥95%

Supplier: AOB CHEM USA
Warning

10159-25G 10159-5G
76906-292EA 488.02 USD InStock
76906-292 76906-294
3-Bromo-2-chloro-4-pyridineboronic acid ≥95%
3-Bromo-2-chloro-4-pyridineboronic acid
Precaución: for Research Use Only


Formula: C₅H₄BBrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD08458470
CAS Number: 1072944-16-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty