Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-methoxyphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

10524-25G 10524-5G
76906-428EA 204.76 USD InStock
76906-428 76906-430
3-Fluoro-5-methoxyphenylboronic acid ≥97%
3-Fluoro-5-methoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD07363780
CAS Number: 609807-25-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty