Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Formyl-5-isopropoxyphenyl)boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

20315-1G 20315-25G 20315-500MG 20315-5G
76910-064EA 162.32 USD InStock
76910-064 76910-066 76910-068 76910-070
(3-Formyl-5-isopropoxyphenyl)boronic acid ≥97% research grade
(3-Formyl-5-isopropoxyphenyl)boronic acid


Formula: C₁₀H₁₃BO₄
Storage Temperature: Ambient
MDL Number: MFCD07784396
CAS Number: 871125-79-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty