Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Fluorophenoxy)-3-methylbenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

23051-1G 23051-25G 23051-500MG 23051-5G
76911-122EA 201.58 USD InStock
76911-122 76911-124 76911-126 76911-128
4-(4-Fluorophenoxy)-3-methylbenzaldehyde ≥97%
4-(4-Fluorophenoxy)-3-methylbenzaldehyde


Formula: C₁₄H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD17304742
CAS Number: 1270878-66-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty