Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,3-dichlorotoluene ≥97% research grade

Supplier: AOB CHEM USA
Warning

31235-1G 31235-25G 31235-500MG 31235-5G
76913-888EA 260.99 USD InStock
76913-888 76913-890 76913-892 76913-894
4-Bromo-2,3-dichlorotoluene ≥97% research grade
4-Bromo-2,3-dichlorotoluene


Formula: C₇H₅BrCl₂
MW: 239.925 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28737861
CAS Number: 1807029-15-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty