Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(3-Pyridyl)aniline ≥97% research grade

Supplier: AOB CHEM USA
Warning

35507-1G 35507-25G 35507-500MG 35507-5G
76915-432EA 171.87 USD InStock
76915-432 76915-434 76915-436 76915-438
4-(3-Pyridyl)aniline ≥97% research grade
4-(3-Pyridyl)aniline


Formula: C₁₁H₁₀N₂
Storage Temperature: Refrigerator
MDL Number: MFCD00956763
CAS Number: 82261-42-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty