Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-methyl-4-pyridinecarboxylic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

20832-1G 20832-500MG 20832-5G
76898-176EA 551.67 USD InStock
76898-176 76898-178 76898-180
2-Fluoro-5-methyl-4-pyridinecarboxylic acid ≥97% research grade
2-Fluoro-5-methyl-4-pyridinecarboxylic acid


Formula: C₇H₆FNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16611273
CAS Number: 1094345-91-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty