Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluoro-5-methoxyphenylboronic acid, pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

31726-1G 31726-500MG 31726-5G
76902-054EA 534.7 USD InStock
76902-054 76902-056 76902-058
2,4-Difluoro-5-methoxyphenylboronic acid, pinacol ester ≥97% research grade
2,4-Difluoro-5-methoxyphenylboronic acid, pinacol ester


Formula: C₁₃H₁₇BF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD12026090
CAS Number: 1150561-57-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty